Learn More
MSC Energizer 321 Silver Oxide Button Cell, Tab Terminal, 100 ea
Catalog No. 5000685282
Encompass
This silver oxide coin and button cell battery is designed for low drain applications in devices such as calculators and watches. It provides a stable 1.55V output and has a capacity of 15 mAh.
  • Disposable battery
  • Size 321
  • Silver oxide chemistry
  • Button tab terminal
  • Disc shape
  • Has a 0.065 in height and 0.268 in diameter
  • Ideal for low drain devices
  • Sold in quantities of 100
Catalog No. 50-006-85282 Supplier MSC Supplier No. 76580901
This silver oxide coin and button cell battery is designed for low drain applications in devices such as calculators and watches. It provides a stable 1.55V output and has a capacity of 15 mAh.
  • Disposable battery
  • Size 321
  • Silver oxide chemistry
  • Button tab terminal
  • Disc shape
  • Has a 0.065 in height and 0.268 in diameter
  • Ideal for low drain devices
  • Sold in quantities of 100