Learn More
MSC Energizer Hearing Aid Battery, Size 312, Zinc-Air, 6 Pack
Catalog No. 5000782221
Encompass
Energizer 312 hearing aid batteries are designed for reliable and long-lasting power in hearing devices. They utilize zinc-air chemistry to provide stable voltage output for optimal performance.
  • Disposable
  • Size 312
  • Zinc-air chemical system
  • Nominal voltage: 1.4 V
  • Typical capacity: 181 mAh
  • Button tab terminal style
  • Compliant with CPSC 16 CFR PART 1700.20
  • Typical weight: 0.5 g
Catalog No. 50-007-82221 Supplier MSC Supplier No. 68045251
Energizer 312 hearing aid batteries are designed for reliable and long-lasting power in hearing devices. They utilize zinc-air chemistry to provide stable voltage output for optimal performance.
  • Disposable
  • Size 312
  • Zinc-air chemical system
  • Nominal voltage: 1.4 V
  • Typical capacity: 181 mAh
  • Button tab terminal style
  • Compliant with CPSC 16 CFR PART 1700.20
  • Typical weight: 0.5 g