Learn More
MSC Gemtor Decelerator II Energy Absorbing Lanyard, 6 ft., 1 3/16 in. Web Width, 1 ea.
Catalog No. 5000685443
Encompass
This energy-absorbing lanyard is designed for fall arrest applications, providing reliable protection for workers at heights. It features a durable design that effectively limits fall arrest forces and is built to meet rigorous safety standards for high-performance fall protection in industrial environments.
  • Provides a maximum fall arrest distance of 42 in.
  • Constructed with a 1 3/16 in. web width
  • Complies with ANSI Z359.13 and A10.32 standards
  • Designed for an 1,800 lb. OSHA capacity
  • Measures 6 ft. in length and contracts to 4.5 ft.
Catalog No. 50-006-85443 Supplier MSC Supplier No. 74108408
This energy-absorbing lanyard is designed for fall arrest applications, providing reliable protection for workers at heights. It features a durable design that effectively limits fall arrest forces and is built to meet rigorous safety standards for high-performance fall protection in industrial environments.
  • Provides a maximum fall arrest distance of 42 in.
  • Constructed with a 1 3/16 in. web width
  • Complies with ANSI Z359.13 and A10.32 standards
  • Designed for an 1,800 lb. OSHA capacity
  • Measures 6 ft. in length and contracts to 4.5 ft.