Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Energy Transformation Balls
GREENER_CHOICESCI_ED

Catalog No. S139423 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139423 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139423 Supplier Supertek™ Supplier No. PH10416
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Demonstrate conversion of kinetic energy into heat and sound energy during a collision when using the Supertek™ Energy Transformation Balls.

  • Consists of a pair of heavy steel balls.
  • When the balls strike against each other with a piece of paper between the balls the heat causes a hole to appear in the paper.
  • The kinetic energy of the balls upon collision is converted to heat which is sufficient to burn a hole in the paper.
  • Instructions included.

Specifications

Grade 6+
Format Kit
Languages English
Subject Physical Science - Force and Mass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.