Learn More
Energy Transformation Kit
SDPSCI_ED
Catalog No. S90537
Change view
Click to view available options
:
Energy Transformation Kit
Catalog No.
S90537 Energy Transformation Kit
1 options
Catalog No. S90537 Supplier Science First Supplier No. 6121315
Includes
Instructions and historical perspective
Convert mechanical energy to heat.
  • Two precision-machined 1 lb. (454g) balls, hardened with chrome-plated steel, demonstrate energy transformation
Certifications
Meets National Science Education Standards and Benchmarks for Science Literacy
Specifications
Product Type Energy Transformation Kit
For Use With (Application) Demonstrates energy transformation, convert mechanical energy to heat
Includes Instructions and historical perspective, two precision-machined 1 lb. balls