Learn More
Yamato Scientific America, Inc. Yamato 90L Forced Convection Oven, Energy saving, 115V
Catalog No. 501898008 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Energy saving, programmable 90L forced convection eco-oven with lower power consumption and CO2 emission. High precision controller allows high performance temperature control and display of CO₂ and power discharge. Program operation with a maximum of 99 steps, 99 patterns repeatable. Standard equipped with various support functions such as calibration offset, power failure recovery mode, save and access of user setting information, as well as other operation modes. Data acquisition from internal test device possible because of cable holes. Easy system upgrade with various option settings. Operating temp ambient +20~210°C.
Catalog No. 50-189-8008 Supplier Yamato Scientific America, Inc. Supplier No. DNE401
Add to Cart
Add to Cart
Request a Quote
Energy saving, programmable 90L forced convection eco-oven with lower power consumption and CO2 emission. High precision controller allows high performance temperature control and display of CO₂ and power discharge. Program operation with a maximum of 99 steps, 99 patterns repeatable. Standard equipped with various support functions such as calibration offset, power failure recovery mode, save and access of user setting information, as well as other operation modes. Data acquisition from internal test device possible because of cable holes. Easy system upgrade with various option settings. Operating temp ambient +20~210°C.