Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Enerisant hydrochloride | 1152749-07-9 | 95.0% | 10 MG
SDP

Catalog No. 5000298435
Encompass_Preferred

Enerisant hydrochloride is the hydrochloride salt of a potent, selective histamine H3 receptor antagonist/inverse agonist supplied as a research reagent for preclinical and in vitro studies. It is provided as a solid for laboratory use and is not intended for human or veterinary applications.

  • Potent histamine H3 receptor antagonist/inverse agonist.
  • Hydrochloride salt form for improved stability and solubility.
  • CAS number 1152749-07-9 and molecular formula C22H31ClN4O3.
  • Molecular weight 434.96 g·mol-1.
  • Typical purity 95.0%.
  • Packaged as a 10 mg solid; store frozen per supplier guidance.

Catalog No. 50-002-98435 Supplier Medchemexpress LLC Supplier No. HY17610A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enerisant hydrochloride is the hydrochloride salt of a potent, selective histamine H3 receptor antagonist/inverse agonist supplied as a research reagent for preclinical and in vitro studies. It is provided as a solid for laboratory use and is not intended for human or veterinary applications.

  • Potent histamine H3 receptor antagonist/inverse agonist.
  • Hydrochloride salt form for improved stability and solubility.
  • CAS number 1152749-07-9 and molecular formula C22H31ClN4O3.
  • Molecular weight 434.96 g·mol-1.
  • Typical purity 95.0%.
  • Packaged as a 10 mg solid; store frozen per supplier guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.