Learn More
MSC Enerpac Hydraulic Bottle Jack, 22 Ton, 5.91 in Stroke, Steel, CE Certified
Catalog No. 5000515905
Encompass
This heavy-duty hydraulic bottle jack offers a 22-ton capacity, 5.91 inch stroke, and 2.95 inch screw extension for industrial and construction lifting. It has a lifting range from 9.21 inches collapsed to 18.07 inches extended. Constructed from robust steel, it meets or exceeds ANSI, PALD, and CE standards for safety and durability.
  • Lower handle effort reduces fatigue.
  • Fully serviceable design with wiper seal ensures extended product life.
  • High-strength beam and pump linkage for extended life.
  • Pumping handle included.
  • Safety relief valve prevents overloading.
  • Automatic by-pass port prevents over-extension.
  • Thick base material with large area for strength and stability.
  • Heat-treated, adjustable extension screw with cleated saddle prevents slipping.
Catalog No. 50-005-15905 Supplier MSC Supplier No. 96107040
This heavy-duty hydraulic bottle jack offers a 22-ton capacity, 5.91 inch stroke, and 2.95 inch screw extension for industrial and construction lifting. It has a lifting range from 9.21 inches collapsed to 18.07 inches extended. Constructed from robust steel, it meets or exceeds ANSI, PALD, and CE standards for safety and durability.
  • Lower handle effort reduces fatigue.
  • Fully serviceable design with wiper seal ensures extended product life.
  • High-strength beam and pump linkage for extended life.
  • Pumping handle included.
  • Safety relief valve prevents overloading.
  • Automatic by-pass port prevents over-extension.
  • Thick base material with large area for strength and stability.
  • Heat-treated, adjustable extension screw with cleated saddle prevents slipping.