Learn More
MSC Enespro Arc Flash Hood, RevoLite, 40 cal/cm2, 1 ea
Catalog No. 5000792631
Encompass
This RevoLite arc flash hood provides inherent flame resistance and enhanced comfort. Designed for high-risk environments, it offers superior protection with an anti-fog faceshield and lighter weight compared to other 40 cal hoods.
  • CAT Level 4 protection
  • Arc rating of 40 cal/cm²
  • Certified NFPA 70E and ASTM F2178
  • Meets ANSI/ISEA Z87.1 standards (Impact Rating Z81+)
  • Fabric: 11.2 oz. RevoLite™
  • Inherently flame resistant; protection will not wash or wear out
  • Anti-fog faceshield with scratch resistant coating
  • PureView faceshield technology for greater visibility
  • Offers 50% lighter weight than other standard 40 cal arc flash PPE
Catalog No. 50-007-92631 Supplier MSC Supplier No. 89948269
This RevoLite arc flash hood provides inherent flame resistance and enhanced comfort. Designed for high-risk environments, it offers superior protection with an anti-fog faceshield and lighter weight compared to other 40 cal hoods.
  • CAT Level 4 protection
  • Arc rating of 40 cal/cm²
  • Certified NFPA 70E and ASTM F2178
  • Meets ANSI/ISEA Z87.1 standards (Impact Rating Z81+)
  • Fabric: 11.2 oz. RevoLite™
  • Inherently flame resistant; protection will not wash or wear out
  • Anti-fog faceshield with scratch resistant coating
  • PureView faceshield technology for greater visibility
  • Offers 50% lighter weight than other standard 40 cal arc flash PPE