Learn More
Description
- Padded handle and shoulder strap
- Large front pocket for electrical glove kit
- Waterproof coating on bottom to keep PPE dry
- U-shaped zipper for easy access to gear
- Padded bag for safety glasses
- Gromments to increase air-flow to suits to control odor
- Custom padded box to protect faceshield
- Care and maintenance printed on the inside bag
Specifications
Specifications
| Product Type | Gear Bag |
| Color | Navy |
| Material | Polyester, PVC |
| Closure Type | Zipper |
| For Use With (Application) | Utilities, Manufacturing, Electrical Contractors |
| Quantity | 1 Ea. |
| Sterility | Non-sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
