Learn More
International Medical Industries (IMI) Tamper Evident Cap for ENFit Syringes (Box of 1000)
Catalog No. NC2030205
Encompass
The Prep-Lock™ Tamper Evident Cap for ENFit™ Syringes is specifically designed to meet the ISO 80369-3 Standard for ENFit Connectors. It is also the only ENFit Syringe Cap available with built-in tamper-evident technology. Compatible with a low dose and standard ENFit Syringes, they provide an additional level of security to syringe contents during transportation, prior to administration. Once the Tamper Evident Cap is installed, it cannot be removed without evidence of access and cannot be reinstalled after removal. Guarding ENFit Syringes with Prep-Lock Tamper Evident Caps easily enhances medication safety protocols and improves patient safety. • Non-Sterile • Box of 1000 • Compatible with Both Standard and Low Dose ENFit Syringes.
Catalog No. NC2030205 Supplier International Medical Industries (IMI) Supplier No. 57220
The Prep-Lock™ Tamper Evident Cap for ENFit™ Syringes is specifically designed to meet the ISO 80369-3 Standard for ENFit Connectors. It is also the only ENFit Syringe Cap available with built-in tamper-evident technology. Compatible with a low dose and standard ENFit Syringes, they provide an additional level of security to syringe contents during transportation, prior to administration. Once the Tamper Evident Cap is installed, it cannot be removed without evidence of access and cannot be reinstalled after removal. Guarding ENFit Syringes with Prep-Lock Tamper Evident Caps easily enhances medication safety protocols and improves patient safety. • Non-Sterile • Box of 1000 • Compatible with Both Standard and Low Dose ENFit Syringes.