Learn More
Selleck Chemical LLC Enfortumab vedotin-ejfv-1MG-5
SDP
Catalog No. 503134772
Encompass_Preferred
Enfortumab vedotin-ejfv (Padcev) is an antibody-drug conjugate (ADC) composed of an fully human anti-Nectin-4 IgG1 kappa monoclonal antibody (AGS-22C3) conjugated to Monomethyl Auristatin E (MMAE) via a protease-cleavable maleimidocaproyl-valyl-citrullinyl-p-aminobenzyloxycarbonyl Enfortumab vedotin-ejfv has the potential for use in research of urothelial carcinoma
Catalog No. 50-313-4772 Supplier Selleck Chemical LLC Supplier No. D40531MG5
Enfortumab vedotin-ejfv (Padcev) is an antibody-drug conjugate (ADC) composed of an fully human anti-Nectin-4 IgG1 kappa monoclonal antibody (AGS-22C3) conjugated to Monomethyl Auristatin E (MMAE) via a protease-cleavable maleimidocaproyl-valyl-citrullinyl-p-aminobenzyloxycarbonyl Enfortumab vedotin-ejfv has the potential for use in research of urothelial carcinoma