Learn More
Cayman Chemical EnfumafungIn 10mg
SDP
Catalog No. 502945924
Encompass_Preferred
A triterpene glycoside with antifungal activity; inhibits 1,3-β-D-glucan synthase in C. albicans (IC50 = 0.2 UG/ml); active against 12 mammalian pathogenic fungi (MICs = ≤0.03 to 2 UG/ml)
Catalog No. 50-294-5924 Supplier Cayman Chemical Supplier No. 3974210 MG
A triterpene glycoside with antifungal activity; inhibits 1,3-β-D-glucan synthase in C. albicans (IC50 = 0.2 UG/ml); active against 12 mammalian pathogenic fungi (MICs = ≤0.03 to 2 UG/ml)