Learn More
Bachem Enfuvirtide Acetate
SDP
Supplier:  Bachem 4062216.0001
Encompass_Preferred
Enfuvirtide Acetate, Ac-Tyr-Thr-Ser-Leu-Ile-His-Ser-Leu-Ile-Glu-Glu-Ser-Gln-Asn-Gln-Gln-Glu-Lys-Asn-Glu-Gln-Glu-Leu-Leu-Glu-Leu-Asp-Lys-Trp-Ala-Ser-Leu-Trp-Asn-Trp-Phe-NH2, 159519-65-0, C204H301N51O64, Mr 4491.94, 1mg. T20, Pentafuside. Enfuvirtide is a 36 amino acid peptide corresponding to a region of gp41, the transmembrane subunit of HIV-1 envelope protein. It belongs to the therapeutic class of fusion inhibitors and acts by binding to gp41 and impeding the conformational changes in gp41 necessary for fusion of the virus with the cell.
Catalog No. 50-383-307