Learn More
TARGETMOL CHEMICALS INC Enfuvirtide acetate 25MG
SDP
Catalog No. 502877338
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Enfuvirtide acetate salt is a linear synthetic peptide made of 36 amino acids. It has an acetylated N-terminus and a carboxamide C-terminus. Purity 99.34%
Catalog No. 50-287-7338 Supplier TARGETMOL CHEMICALS INC Supplier No. TP143225MG
Also available in 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Enfuvirtide acetate salt is a linear synthetic peptide made of 36 amino acids. It has an acetylated N-terminus and a carboxamide C-terminus. Purity 99.34%