Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Acid-Fast Stain Kit, Modification of the Kinyoun Method

Catalog No. ES6500
Change view
Click to view available options
Stain Type:
Brilliant Green
Loeffler's Methylene Blue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stain Type
ES6500 Loeffler's Methylene Blue
ES6501 Brilliant Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. ES6500 Supplier ENG Scientific Supplier No. 6500
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Acid-Fast Stain Kit, Modification of the Kinyoun Method, is ideal for staining acid-fast bacteria. Use with Acid-fast stain kit.

  • Modification of the Kinyoun Method
  • Acid-fast bacilli stain red; all other bacteria stain blue with Sol. III, green with Sol. IIIA.
  • Acid Fast Stain Kit
  • For in vitro diagnostic use
  • 1 Kit

Specifications

Quantity 1 Ea.
Includes 250 mL carbol fuchsin, 250 mL acid alcohol and 250 mL Loeffler's methylene blue
For Use With (Application) In vitro diagnostic use
Color Dark Red
Stain Type Loeffler's Methylene Blue
Volume (Metric) 250 mL
Form Liquid
Product Type Acid Fast Stain
Packaging Kit

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.