Learn More
ENG Scientific Albert's Stain Solution I
Catalog No. ES6760
Change view
Click to view available options
Quantity:
2 x 100 mL
Catalog No. Quantity
ES6760 2 x 100 mL
1 options
Catalog No. ES6760 Supplier ENG Scientific Supplier No. 6760
Add to Cart
Add to Cart
Albert's Stain Solution I for Diptheria Bacilli. Differentiates volutin granules in cells and reveals barred staining patterns of cytoplasmic regions. Includes Solutions I and II.
  • Albert's Stain Solution I
  • Replacement Solution for Albert's Stain Solution Kit
  • For staining Diptheria bacilli
Specifications
For Use With (Application) Staining Diptheria bacilli
Packaging Bottles
Quantity 2 x 100 mL
Product Type Replacement Solution for Albert's Stain Solution Kit
For in vitro diagnostic use only.