Learn More
ENG Scientific Albert's Stain Solution II
Catalog No. ES6770
Change view
Click to view available options
Quantity:
2 x 100 mL
Catalog No. Quantity
ES6770 2 x 100 mL
1 options
Catalog No. ES6770 Supplier ENG Scientific Supplier No. 6770
Add to Cart
Add to Cart
Albert's Stain Solution II for Diptheria Bacilli. Differentiates volutin granules in cells and reveals barred staining patterns of cytoplasmic regions. Includes Solutions I and II.
  • Albert's Stain Solution II
  • Replacement Solution for Albert's Stain Solution Kit
  • For staining Diptheria bacilli
Specifications
For Use With (Application) Staining Diptheria bacilli
Packaging Bottles
Quantity 2 x 100 mL
Product Type Replacement Solution for Albert's Stain Solution Kit
For in vitro diagnostic use only.