Learn More
ENG Scientific Albert's Stain Solution Kit
Catalog No. ES6750
Change view
Click to view available options
Quantity:
2 x 100 mL
Catalog No. Quantity
ES6750 2 x 100 mL
1 options
This item is not returnable. View return policy
Catalog No. ES6750 Supplier ENG Scientific Supplier No. 6750
Add to Cart
Add to Cart
This item is not returnable. View return policy
Albert's Stain Solution Kit for Diptheria Bacilli. Differentiates volutin granules in cells and reveals barred staining patterns of cytoplasmic regions. Includes Solutions I and II.
  • This method differentiates volutin granules in the cells, and reveals the barred staining patterns of the cytoplasmic region.
  • Recommended Procedure:
    • Stain heat-fixed smear with Sol. I for 3 - 5 minutes.
    • Wash in water, blot and air dry.
    • Stain with Sol. II for 1 minute.
    • Wash again, blot and air dry.
  • Results: Volutin granules - dark blue or black; cytoplasm-light green; barred patterns - dark green.
  • Contents: Toluidine Blue, Malachite Green, Ethanol, Acetic Acid, Iodine, Potassium Iodide.
  • These solutions are made from certified dyes (when applicable).
  • Albert's Stain Solution Kit
  • Includes 100 mL Albert's stain solution I and 100 mL Albert's stain solution II
  • For in vitro diagnostic use
  • 1 Kit
Specifications
Quantity 2 x 100 mL
Includes 100 mL Albert's stain solution I and 100 mL Albert's stain solution II
For Use With (Application) In vitro diagnostic use
Volume (Metric) 100 mL
Product Type Albert's Stain Solution Kit
Packaging Complete Kit (Bottles)
For in vitro diagnostic use only.