Learn More
ENG Scientific Bone Marrow Stain Kit for Iron, Solution II - Hydrochloric Acid (1%)
Description
- 1% Hydochloric Acid, Solution II
- Bone Marrow Stain Kit For Iron
- For Iron (Prussian blue reaction)
Specifications
Specifications
| Quantity | 1 x 3800 mL |
| For Use With (Application) | For Iron (Prussian blue reaction) |
| Packaging | Bottle |
| Product Type | Reagent |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.