Learn More
ENG Scientific Traxstain Buffer Solution
Description
- Buffer Solution Traxstain
- For EM Sciences Midas™ slide stainers, Geometric Data Corporation Hemastainer™ or any other instrument employing a dip stain technique
- For in-vitro diagnostic use only
Specifications
Specifications
| Quantity | 1 x 19 L |
| For Use With (Application) | For in-vitro diagnostic use only |
| For Use With (Equipment) | For EM Sciences Midas™ slide stainers, Geometric Data Corporation Hemastainer™ or any other instrument employing a dip stain technique |
| pH | 6.75 |
| Packaging | Cubetainer |
| Product Type | Staining Buffer |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.