Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Buffered Neutral Formalin (10%)

Catalog No. ES9700
Change view
Click to view available options
Quantity:
1 x 19 L (5 gal.)
2 x 950 mL
Packaging Type:
Bottle
Cubetainer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
ES9700 2 x 950 mL Bottle
ES9720 1 x 19 L (5 gal.) Cubetainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. ES9700 Supplier ENG Scientific Supplier No. 9700
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Buffered Neutral Formalin (10%)

  • Buffered Neutral Formalin (10%)

Specifications

Concentration 10%
Type Buffered Neutral Formalin (10%)
Packaging Type Bottle
Quantity 2 x 950 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.