Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Calcofluor White Stain, Certified

Catalog No. ES6726
Change view
Click to view available options
Quantity:
100 mL
2 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6726 100 mL
ES6725 2 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. ES6726 Supplier ENG Scientific Supplier No. 6726
Add to Cart
Add to Cart
This item is not returnable. View return policy

Calcofluor white, a nonspecific fluorochrome, will bind to the cell walls of certain fungi allowing for the direct microscopic observation of fungi in wet preparations.

  • Fungi elements fluoresce blue-white or green depending on filter system used
  • A G-365 excitation filter and an LP 420 barrier filter produce light of blue-white wavelength
  • A K530 excitation filter and a BG 12 barrier filter produce light of green wavelength
  • Calcofluor White Stain
  • Includes Evans blue
  • For in vitro diagnostic use

Specifications

Quantity 100 mL
Includes Evans blue
Format Bottle
For Use With (Application) In vitro diagnostic use
Stain Type Calcofluor White
Volume (Metric) 100 mL
Product Type Calcofluor White Stain

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.