Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Calcofluor White Stain, Certified

Catalog No. p-3201185
Click to view available options
Quantity:
100 mL
2 x 100 mL
This item is not returnable. View return policy

Catalog No. ES6726

Supplier: ENG Scientific 6726

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Calcofluor white, a nonspecific fluorochrome, will bind to the cell walls of certain fungi allowing for the direct microscopic observation of fungi in wet preparations.

  • Fungi elements fluoresce blue-white or green depending on filter system used
  • A G-365 excitation filter and an LP 420 barrier filter produce light of blue-white wavelength
  • A K530 excitation filter and a BG 12 barrier filter produce light of green wavelength
  • Calcofluor White Stain
  • Includes Evans blue
  • For in vitro diagnostic use

Specifications

Type Calcofluor White Stain
Quantity 100 mL
Includes Evans blue
Format Bottle
For Use With (Application) In vitro diagnostic use
Stain Type Calcofluor White

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.