Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Chlamydial Stain Kit

Catalog No. ES7100
Change view
Click to view available options
Quantity:
Solution I: 1 x 250 mL, Solution II: 2 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES7100 Solution I: 1 x 250 mL, Solution II: 2 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES7100 Supplier ENG Scientific Supplier No. 7100
Add to Cart
Add to Cart

The Chlamydial Stain Kit includes 250 mL Solution I - Giemsa Stain, 100 mL Solution II - Buffer solution and 100 mL Solution III - Iodine Stain.

  • Kit Includes Solution I - Giemsa Stain 250 mL, Solution II - Buffer Solution 100 mL, and Solution III - Iodine Stain 100 mL
  • Recommended Procedure:
    • Giemsa Buffer Solution:
      • 1 part of Sol. I to 40 or 50 parts of Sol. II. (If a darker stain is desired, increase ratio of stain to buffer.)
      • Place fixed smear into Giemsa Buffer Sol. for 1 hour. (Time may vary according to intensity of stain desired.)
      • NOTE: Giemsa Buffer Sol. should be freshly made.
      • Slide is then rinsed rapidly in 95% ethanol to remove excess dye and to enhance differentiation.
    • Iodine Method:
      • Place fixed smear into Sol. III for 3 - 5 minutes.
      • Slide is examined as a wet mount.
      • NOTE: The slide may be decolorized with methyl alcohol and restained with Giemsa stain.
    • Results:
      • Giemsa Stain:
        • The elementary bodies stain a reddish purple.
        • The initial bodies are more basophilic, staining bluish as do bacteria.
      • Iodine Stain:
        • The matrix of inclusions may appear as a reddish-brown mass recognizable under low power.
        • Check with higher objective to confirm.
  • Contents: Giemsa Stain, Methanol, Buffer Salts, Iodine, Potassium Iodide
  • These solutions are made from certified dyes (when applicable).
  • For in vitro diagnostic use
  • Chlamydial Stain Kit
  • 1 Kit

Specifications

Quantity Solution I: 1 x 250 mL, Solution II: 2 x 100 mL
Includes 250 mL Solution I - Giemsa stain, 100 mL Solution II - buffer Solution and Solution III - iodine stain
For Use With (Application) In vitro diagnostic use
Product Type Chlamydial Stain Kit
Packaging Complete Kit (Bottles)

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.