Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Cryptosporidium Stain Kit

Catalog No. ES6470
Change view
Click to view available options
Quantity:
2 x 250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6470 2 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ES6470 Supplier ENG Scientific Supplier No. 6470
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentiates Cryptosporidium oocysts from yeast cells. Specimens may be prepared from fresh material or preserved in 10% buffered formalin. For use with Cryptosporidium Stain Kit (Modified Kinyoun method).

  • Fresh specimens should be fixed in methanol and formalized specimens with heat
  • Ideal for differential staining of bacteria
  • Yeast cells will appear green to blue-purple
  • Decolorization and counterstaining times may vary according to thickness of specimen
  • Cryptosporidium oocysts will appear light pink to deep red
  • Utilizes the Modified Kinyoun Method, which aids in differentiating Crytosporidium oocysts from yeast cells
  • Cryptosporidium Stain Kit
  • Includes Solution I - Carbol Fuchsin 250 mL, Solution II - Acid Alcohol 250 mL, Solution III - Concentrated Light Green 250 mL
  • For in vitro diagnostic use
  • 1 Kit

Specifications

Quantity 2 x 250 mL
Includes 250 mL Solution I - carbol fuchsin, 250 mL Solution II - acid alcohol and 250 mL Solution III - concentrated light green
For Use With (Application) In vitro diagnostic use
Stain Type Cryptosporidium
Volume (Metric) 250 mL
Product Type Cryptosporidium Stain Kit
Packaging Complete Kit (Bottles)

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.