Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Cyclospora Stain Kit (Modified Kinyoun Method)

Catalog No. ES6465
Change view
Click to view available options
Quantity:
3 x 250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6465 3 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES6465 Supplier ENG Scientific Supplier No. 6465
Add to Cart
Add to Cart

The Cyclospora Stain Kit includes Solution I - Carbol Fuchsin, Solution II - Acid Alcohol (10% Sulfuric), and Solution III - Malachite Green.

  • Cyclospora Stain Kit
  • Includes Solution I - Carbol Fuchsin, Solution II - Acid Alcohol (10% Sulfuric), Solution III - Malachite Green
  • For staining Cyclospora oocysts

Specifications

Quantity 3 x 250 mL
Includes Solution I - Carbol Fuchsin; Solution II - Acid Alcohol (10% Sulfuric); Solution III - Malachite Green
For Use With (Application) Staining Cyclospora oocysts
Volume (Metric) 250 mL

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.