Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Diazo Method Stain Kit

Catalog No. ES9520
Change view
Click to view available options
Quantity:
3 x 250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES9520 3 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES9520 Supplier ENG Scientific Supplier No. 9520
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diazo Method Stain Kit for Argentaffin granules. Includes Brentamine Fast Red B, Lithium Carbonate, Hematoxylin, Citric Acid, Sodium Iodate, Potassium Alum.

  • Diazo Method Stain Kit
  • Made from certified dyes (when applicable)
  • Kit includes:
    • Solution I - Brentamine Fast Red B (1% Stock)
    • Solution II - Lithium Carbonate (Stock)
    • Solution III - Mayer's Acid Alum Hematoxylin
  • 1 Kit

Specifications

For Use With (Application) Argentaffin granules
Includes Brentamine Fast Red B, Lithium Carbonate, Hematoxylin, Citric Acid, Sodium Iodate, Potassium Alum
Packaging Complete Kit (Bottles)
Quantity 3 x 250 mL
Solution Type Solutions I - III
Product Type Diazo Method Stain Kit

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.