Learn More
ENG Scientific Eosinophil Diluting Fluid and Stain
Catalog No. ES3100
Change view
Click to view available options
Quantity:
2 x 100 mL
Catalog No. Quantity
ES3100 2 x 100 mL
1 options
Catalog No. ES3100 Supplier ENG Scientific Supplier No. 3100
Add to Cart
Add to Cart
Eosinophil Diluting Fluid and Stain is useful in determining an increase (eosinophilia) or decrease (eosinopenia) in eosinophilic granulocytes. Requires special diluent and/or stain solution.
  • Diluting Fluid and Stain Eosinophil
Specifications
Quantity 2 x 100 mL
Packaging Bottles
Product Type Stain
For in vitro diagnostic use only.