Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Dip Stain Kit (Wright and Wright Giemsa) Components

Catalog No. ES2590
Change view
Click to view available options
Product Type:
Wright Dip Stain - Solution II
Wright Giemsa Dip Stain - Solution II
Wright/Wright Giemsa Dip Stain - Solution I
Wright/Wright Giemsa Dip Stain - Solution IV
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
ES2590 Wright/Wright Giemsa Dip Stain - Solution IV
ES2570 Wright Giemsa Dip Stain - Solution II
ES2560 Wright Dip Stain - Solution II
ES2550 Wright/Wright Giemsa Dip Stain - Solution I
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ES2590 Supplier ENG Scientific Supplier No. 2590
Add to Cart
Add to Cart

For Hemotology staining using Dip Stain Kit (Wright & Wright Giemsa). Kit includes Solution I - Fixative, Solution II - Wright or Wright Giemsa Dip Stain, Solution III - Matching Buffer, and Solution IV - Alcohol Wash. Solutions made from certified dyes.

  • Solutions made from certified dyes
  • Solutions should be replenished during staining period
  • During cold weather, allow solutions to return to room temperature and shake well before use

Specifications

Quantity 3800 mL (1 gal.)
Storage Requirements Room Temperature
For Use With (Application) For in-vitro diagnostic use only
Product Type Wright/Wright Giemsa Dip Stain - Solution IV

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.