Learn More
ENG Scientific Fecal Leukocyte Stain (Loeffler' Methylene Blue)
Description
- Contents: Ethanol, Methylene Blue
- This solution is made from certified dyes.
Specifications
Specifications
| Quantity | 2 x 100 mL |
| For Use With (Application) | For microscopic identification and counting of fecal leukocytes |
| Staining Time | 2 to 3 min. |
| Packaging | Bottles |
| Product Type | Stain |
For in vitro diagnostic use only.