Learn More
ENG Scientific Fite's Method Stain Kit
Catalog No. ES9580
Change view
Click to view available options
Quantity:
3 x 250 mL
Catalog No. Quantity
ES9580 3 x 250 mL
1 options
Catalog No. ES9580 Supplier ENG Scientific Supplier No. 9580
Add to Cart
Add to Cart
Fite's Method Stain Kit for visualizing acid-fast bacteria in tissue.
  • Fite's Method Stain Kit
  • Includes Basic Fuchsin, Phenol, Ethanol, Sulfuric Acid, Methylene Blue
  • Visualizes acid-fast bacteria in tissue
  • 1 Kit
Specifications
For Use With (Application) Visualizes acid-fast bacteria in tissue
Includes Basic Fuchsin, Phenol, Ethanol, Sulfuric Acid, Methylene Blue
Packaging Complete Kit (Bottles)
Quantity 3 x 250 mL
Product Type Fite's Method Stain Kit
For in vitro diagnostic use only.