Learn More
ENG Scientific Fixative Traxstain
Description
- In the event of cold weather, allow solution to return to room temperature and shake well before use.
- Contents: Wright Stain, Giemsa Stain, May-Grunwald Stain, Methanol, Buffers, Stabilizers.
- These solutions are made from certified dyes (when applicable).
Specifications
Specifications
| Quantity | 1 x 19 L (5 gal.) |
| Storage Requirements | Room Temperature |
| For Use With (Application) | For in-vitro diagnostic use only |
| For Use With (Equipment) | For EM Sciences Midas™ slide stainers, Geometric Data Corporation Hemastainer™ or any other instrument employing a dip stain technique |
| Packaging | Cubetainer |
| Product Type | Fixative Solution |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.