Learn More
ENG Scientific Fluorescent Stain Kit
Description
- Acid-fast organisms should appear fluoresced against a dark background
- Fluorescent Stain Kit
- Includes 250 mL auramine phenol solution, 250 mL acid alcohol, and 250 mL potassium permanganate
- In vitro diagnostic use
Specifications
Specifications
| Quantity | 3 x 250 mL |
| Includes | 250 mL auramine phenol solution, 250 mL acid alcohol and 250 mL potassium permanganate |
| For Use With (Application) | In vitro diagnostic use |
| Stain Type | Fluorescent |
| Product Type | Fluorescent Stain Kit |
| Packaging | Complete Kit (Bottles) |
For in vitro diagnostic use only.