Learn More
ENG Scientific Gomori's Trichrome Stain, Solution III - Aqueous Safranin (0.1%)
Description
- Gomori's Trichrome Stain
- Aqueous Safranin Solution IIIA
- Test time 3- 5 minutes
- For in vitro diagnostic use only
Specifications
Specifications
| Packaging Type | Bottle |
| Concentration | 0.1% |
| For Use With (Equipment) | Staining Equipment |
| Product Type | Staining Reagent |
| Quantity | 1 x 250 mL |
| Solution Type | Stain |
| For Use With (Application) | In vitro diagnostic use only |
| Test Time | 3 to 5 min. |
| Form | Solution |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.