Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Gomori's Trichrome Stain, Hydrochloric Acid (20%), Solution I

Catalog No. ES9141
Change view
Click to view available options
Quantity:
1 x 250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES9141 1 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES9141 Supplier ENG Scientific Supplier No. 9141
Add to Cart
Add to Cart

Gomori's, Trichrome Stain, Hydrochloric Acid (20%), Solution I is designed to demonstrate deposits of hemosiderin (iron) in tissue specimens.

  • Gomori's Trichrome Stain
  • ; Hydrochloric Acid (20%), Solution I
  • For in vitro diagnostic use only

Specifications

Packaging Type Bottle
For Use With (Equipment) Staining Equipment
Product Type Hydrochloric Acid Solution
Quantity 1 x 250 mL
Solution Type Buffer
For Use With (Application) In vitro diagnostic use only
Form Solution

For in vitro diagnostic use only. Due to product restrictions, please Sign In to purchase or view availability for this product.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.