Learn More
ENG Scientific Gomori’s Method for Iron in Tissue with Solution IIIA Safranin
Description
- Nuclear Fast Red Solution IIIB
- In vitro diagnostic use only
- Test time 3 to 5 min.
Specifications
Specifications
| Packaging Type | Bottles |
| For Use With (Equipment) | Staining Equipment |
| Product Type | Staining Reagent |
| Quantity | 3 x 250 mL |
| For Use With (Application) | In vitro diagnostic use only |
| Test Time | 3 to 5 min. |
| Form | Solution |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.