Learn More
ENG Scientific Gomori's Trichrome Stain, Potassium Ferrocyanide (10%), Solution II
Description
- Gomori's Trichrome Stain
- Potassium Ferrocyanide Solution II
- For in vitro diagnostic use only
Specifications
Specifications
| Packaging Type | Bottle |
| For Use With (Equipment) | Staining Equipment |
| Product Type | Staining Reagent |
| Quantity | 1 x 250 mL |
| Solution Type | Stain |
| For Use With (Application) | In vitro diagnostic use only |
| Form | Solution |
For in vitro diagnostic use only. Due to product restrictions, please Sign In to purchase or view availability for this product.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.