Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Gomori's Trichrome Stain

Catalog No. ES9000
Change view
Click to view available options
Quantity:
2 x 950 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES9000 2 x 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES9000 Supplier ENG Scientific Supplier No. 9000
Add to Cart
Add to Cart

Gomori's Trichrome Stain demonstrates fine histological detail of connective tissue, collagen and muscle fibers.

  • Gomori's Trichrome Stain
  • Includes Chromotrope 2R, Light Green SF, Glacial Acetic Acid, Phosphotungstic Acid
  • For in vitro diagnostic use only

Specifications

Content And Storage Chromotrope 2R, Light Green SF, Glacial Acetic Acid, Phosphotungstic Acid
Packaging Type Bottles
For Use With (Equipment) Staining Equipment
Product Type Staining Reagent
Quantity 2 x 950 mL
Solution Type Stain
For Use With (Application) In vitro diagnostic use only
Form Liquid

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.