Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Gram Stain Kit with Carbol Fuchsin Counterstain

Catalog No. ES9579
Change view
Click to view available options
Quantity:
1 x 250 mL
4 x 250 mL
Product Type:
Carbol Fuchsin
Gram Stain Kit
Packaging:
Bottle
Complete Kit (Bottles)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Packaging
ES9579 1 x 250 mL Carbol Fuchsin Bottle
ES9575 4 x 250 mL Gram Stain Kit Complete Kit (Bottles)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES9579 Supplier ENG Scientific Supplier No. 9579
Add to Cart
Add to Cart

For bacteria in tissue. Kit and individual components available for Gram Stain Kit - For Bacteria in Tissue with Carbol Fuchsin Counterstain. Contains crystal violet, iodine, potassium oxalate, ethanol, acetone, safranin, basic fuchsin, and phenol.

  • Contains crystal violet, iodine, potassium oxalate, ethanol, acetone, safranin, basic fuchsin, phenol
  • Gram-positive bacteria, many fungi and fibrin stain blue-black to violet
  • Gram-negative bacteria stain red
  • Cytoplasm stains pink

Specifications

Quantity 1 x 250 mL
For Use With (Application) In vitro diagnostic use
Stain Type Gram Stain
Volume (Metric) 250 mL
Product Type Carbol Fuchsin
Packaging Bottle

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.