Learn More
ENG Scientific Gram Stain Kit with Safranin Counterstain
Description
- Contains crystal violet, iodine, potassium oxalate, ethanol, acetone, safranin, basic fuchsin, phenol
- Gram-positive bacteria, many fungi and fibrin stain blue-black to violet
- Gram-negative bacteria stain red
- Cytoplasm stains pink
Specifications
Specifications
| Quantity | 4 x 250 mL |
| For Use With (Application) | In vitro diagnostic use |
| Stain Type | Gram Stain |
| Volume (Metric) | 250 mL |
| Product Type | Crystal Violet, Safranin |
| Packaging | Complete Kit (Bottles) |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.