Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Gram Stain Kit with Safranin Counterstain

Catalog No. ES9574
Change view
Click to view available options
Product Type:
Crystal Violet
Crystal Violet, Safranin
Gram Iodine
Safranin
For Use With (Application):
Bacteria in tissue
Gram staining
In vitro diagnostic use
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Application)
ES9574 Safranin Gram staining
ES9570 Crystal Violet, Safranin In vitro diagnostic use
ES9571 Crystal Violet Bacteria in tissue
ES9572 Gram Iodine Bacteria in tissue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ES9574 Supplier ENG Scientific Supplier No. 9574
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For bacteria in tissue. Kit and individual components available for Gram Stain Kit - Bacteria in Tissue w/ Safaranin Counterstain. Contains crystal violet, iodine, potassium oxalate, ethanol, acetone, safranin, basic fuchsin, and phenol.

  • Contains crystal violet, iodine, potassium oxalate, ethanol, acetone, safranin, basic fuchsin, phenol
  • Gram-positive bacteria, many fungi and fibrin stain blue-black to violet
  • Gram-negative bacteria stain red
  • Cytoplasm stains pink

Specifications

Quantity 1 x 250 mL
For Use With (Application) Gram staining
Stain Type Gram Stain
Volume (Metric) 250 mL
Product Type Safranin
Packaging Bottle

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.