Learn More
ENG Scientific H-Pack™ II Wright Giemsa Stain
Description
- One adjustment should be all that is necessary to obtain desired color and intensity
- Buffer solution compliments individual stain lots to allow consistent staining characteristics
- Normal and abnormal WBC's and platelets are vividly demonstrated
- Bacteria within leukocytes can be clearly observed
- RBC staining gives excellent results of normal cellular elements including all abnormalities and inclusion bodies
- Parasites, such as malaria, are well-defined. Bone marrow preps can be stained routinely
- Freezing or high temperature will not cause change in staining ability
- Stain pack is designed to be used at room temperature
- SHAKE WELL BEFORE USE
- Designed to eliminate problems with precipitate, artifacts, drying of slides and overstaining
- Use with Ames Hema-Tek™ II slide stainers
Specifications
Specifications
| Quantity | 1 Cs. |
| Storage Requirements | Room Temperature |
| For Use With (Application) | Designed to eliminate problems with precipitate, artifacts, drying of slides and overstaining |
| For Use With (Equipment) | For Ames Hema-Tek™ II slide stainers |
| Product Type | Stain |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.