Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific H-Pack™ May-Grunwald Giemsa Stain Packs

Catalog No. ES1150
Change view
Click to view available options
Quantity:
1 Pk.
6 Pk./Cs., 1 Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES1150 6 Pk./Cs., 1 Cs.
ES1151 1 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
The solvent for the stain is methanol, which is flammable.
Catalog No. ES1150 Supplier ENG Scientific Supplier No. 1150
Add to Cart
Add to Cart
The solvent for the stain is methanol, which is flammable.

For Ames Hema-Tek™ slide stainers. Designed to eliminate problems with precipitate, artifacts, drying of slides, and overstaining which causes abnormal staining characteristics of all cellular elements, i.e., false toxic granulation.

  • For AMES HEMA-TEK™ slide stainers
  • To obtain desired color and intensity
  • To demonstrate bacteria within leukocytes, normal and abnormal white blood cells and platelets clearly
  • Freezing or high temperature does not affect staining ability
  • Designed to be used at room temperature
  • Offered with matched buffer solution of specific pH
  • To stain LE preps, parasitic elements like malaria, thicker buffy coat smears, mature and immature elements of routine bone marrow smears, inclusion bodies like basophilic stippling

Specifications

Quantity 6 Pk./Cs., 1 Cs.
Storage Requirements Room Temperature
For Use With (Application) Designed to eliminate problems with precipitate, artifacts, drying of slides and overstaining
For Use With (Equipment) For Ames Hema-Tek™ II slide stainers
Product Type Stain Pack

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.