Learn More
ENG Scientific H-Pack™ Wright Giemsa Stains
Description
- Hematology stain formulated to minimize false toxic granulation
- Designed to eliminate problems with precipitate, artifacts, drying of slides, and overstaining
- High temperature will not cause change in staining ability
- Methanol, buffers and stabilizers required for staining procedure
- Buffer solution complements individual stain lots to allow consistent staining
- Solutions made from certified dyes
- During cold weather, allow solutions to return to room temperature and shake well before use
Used on all specimens requiring WBC differential, particularly spinal fluid and nasal smears, and any procedure requiring Wright counterstain. Other uses include LE preps, parasitic elements such as malaria, thicker buffy coat smears, mature and immature elements of routine bone marrow smears, inclusion bodies such as basophilic stippling.
Specifications
Specifications
| Quantity | 6 Pk./Cs., 1 Cs. |
| Storage Requirements | Room Temperature |
| For Use With (Equipment) | For Ames Hema-Tek™ slide stainers |
| Packaging | H-Pack |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.