Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eng Scientific H-Pack™ Wright Stains

Catalog No. ES1001
Change view
Click to view available options
Quantity:
1 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES1001 1 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ES1001 Supplier Eng Scientific Supplier No. 1001
This item is not returnable. View return policy

For Ames Hema-Tek™ slide stainers

  • Hematology stain formulated to minimize false toxic granulation
  • Designed to eliminate problems with precipitate, artifacts, drying of slides, and overstaining
  • High temperature will not cause change in staining ability
  • Methanol, buffers and stabilizers required for staining procedure
  • Buffer solution complements individual stain lots to allow consistent staining
  • Solutions made from certified dyes
  • During cold weather, allow solutions to return to room temperature and shake well before use

Used on all specimens requiring WBC differential, particularly spinal fluid and nasal smears, and any procedure requiring Wright counterstain. Other uses include LE preps, parasitic elements such as malaria, thicker buffy coat smears, mature and immature elements of routine bone marrow smears, inclusion bodies such as basophilic stippling.

Specifications

Quantity 1 Pk.
Storage Requirements Room Temperature
For Use With (Application) For use in visualizing normal and abnormal WBCs and platelets, Parasites and Bacteria within WBCs from many kinds of samples including CSF, nasal smears and bone marrow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.