Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Heidenhain's Iron Hematoxylin Stain Kit

Catalog No. ES8702
Change view
Click to view available options
Quantity:
2 x 250 mL
2 x 3.8 L (1 gal.)
2 x 950 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES8702 2 x 3.8 L (1 gal.)
ES8700 2 x 250 mL
ES8701 2 x 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ES8702 Supplier ENG Scientific Supplier No. 8702
Add to Cart
Add to Cart

Includes

Made from certified dyes. Kit includes: Solution I - Ammonio-Ferric Alum and Solution II - 0.5% aqueous Hematoxylin.

Eliminates artificial or natural ripening. For dense, regressive staining.

  • A histologic stain composed of ferric salts which ripen and oxidize hematoxylin rapidly
  • Solutions made with iron salts are useful for dense regressive staining
  • Differentiation can be accurately controlled, and the staining is consistent
  • Made from certified dyes
  • Kit includes: Solution I - Ammonio-Ferric Alum and Solution II - 0.5% aqueous Hematoxylin

Specifications

For Use With (Application) Used for dense regressive staining
Includes Ferric Ammonium Sulfate, Hematoxylin
Packaging Kit (Bottles)
Quantity 2 x 3.8 L (1 gal.)
Product Type Heidenhain's Iron Hematoxylin Stain Kit

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.