Learn More
ENG Scientific Heinz Body Stain
Catalog No. ES5501
Change view
Click to view available options
Quantity:
100 mL
2 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES5501 100 mL
ES5500 2 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. ES5501 Supplier ENG Scientific Supplier No. 5501
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use this helpful Heinz Body Stain to recognize hemolytic anemia due to certain drugs and in detecting red cells of the primaquine-sensitive type by detecting Heinz bodies.
  • Heinz bodies are best stained supravitally
  • Includes Methyl Violet, Saline (NaCl)
  • Made from certified dyes
Specifications
Quantity 100 mL
For Use With (Application) Hematology Staining
Packaging Bottle
Product Type Heinz Body Stain
For in vitro diagnostic use only.