Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Lactophenol Cotton Blue Stain, Certified

Catalog No. ES6731
Change view
Click to view available options
Quantity:
1 x 100 mL
2 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6731 1 x 100 mL
ES6730 2 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES6731 Supplier ENG Scientific Supplier No. 6731
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

For staining fungi using Lactophenol Cotton Blue Stain.

  • Lactic acid acts as a preservative for fungi, killed by phenol and stained by cotton blue
  • Fungal elements are stained a deep blue; background is pale blue
  • Lactophenol Cotton Blue Stain
  • Includes Phenol, lactic acid, glycerol, aniline blue
  • For use with staining fungi, in-vitro diagnostic use/li>

Specifications

Quantity 1 x 100 mL
Includes Phenol, lactic acid, glycerol, aniline blue
Format Bottle
For Use With (Application) Staining fungi, in-vitro diagnostic use
Stain Type Lactophenol Cotton Blue
Volume (Metric) 100 mL
Product Type Lactophenol Cotton Blue Stain

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.