Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Leifson Bacterial Flagella Stain Kit

Catalog No. ES6980
Change view
Click to view available options
Quantity:
4 x 250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6980 4 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES6980 Supplier ENG Scientific Supplier No. 6980
Add to Cart
Add to Cart

Leifson Bacterial Flagella Stain Kit is used to establish polar or lateral distribution of flagella on a bacterial cell. Includes Solution I - Basic Fuchsin, (1.2%) Alcoholic, Solution II - Tannic Acid (3%), Solution III - Sodium chloride (1.5%), and Solution IV - Methylene Blue.

  • Kit Includes Solution I - Basic Fuchsin, (1.2%) Alcoholic 250 mL, Solution II - Tannic Acid (3%) 250 mL, Solution III - Sodium Chloride (1.5%) 250 mL, and Solution IV - Methylene Blue 250 mL
  • Bacterial flagella are below the visual limit in size, approximately 0.1 to 0.3 μ in diameter.
  • Flagella stains increase the width of these delicate appendages by forming a precipitate at the surface of the flagella making them visible with the optical microscope.
  • The preparation may be used to establish polar or lateral distribution of flagella on a bacterial cell.
  • Recommended Procedure:
    • Preparation of Slides:
      • Slides must be clean and grease free.
      • To assure this, soak slides for 4 days in a slide cleaning solution (Product No. 6981).
      • Thoroughly wash slides in distilled water and allow to dry.
    • Working Solution:
      • Mix equal parts of each solution (I, II, III) in a glass vessel and tightly stopper.
      • Allow to stand 2 hours prior to use.
      • NOTE: Working stain solution will stain flagella satisfactorily for only 2 - 3 days; therefore, only prepare as needed.
      • This solution precipitates rapidly at room temperature.
    • Use only the clear supernatant for staining.
    • From a centrifuged 24 hour broth culture of a motile species of bacteria, a loopful of sediment is placed at the end of the slide.
    • Tilt the slide allowing the liquid to run slowly to the other end and air dry.
    • Quickly add 1 mL of stain mixture.
    • Allow to stand 5 - 15 minutes at room temperature.
    • When precipitate forms over the entire area of the slide, carefully wash off by flooding with water.
    • Counterstain with 1:10 dilution of Sol. IV for 1 minute.
    • Wash and drain dry.
  • Results: Flagella stain bright red.
  • Contents: Basic Fuchsin, Ethanol, Tannic Acid, Sodium Chloride, Methylene Blue, HCl
  • These solutions are made from certified dyes (when applicable).
  • For in vitro diagnostic use.
  • 1 Kit

Specifications

Quantity 4 x 250 mL
Includes 250 mL Solution I - basic fuchsin, 250 mL (1.2%) alcoholic, 250 mL Solution II - tannic acid (3%), 250 mL Solution III - sodium chloride (1.5%), 250 mL Solution IV - methylene blue
For Use With (Application) In vitro diagnostic use
Volume (Metric) 250 mL
Product Type Leifson's Bacterial Flagella Stain Kit
Packaging Complete Kit (Bottles)

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.